Learn More
Biperiden.HCl, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Biperiden.HCl
Specifications
Specifications
| Chemical Name or Material | Biperiden hydrochloride |
| CAS | 1235-82-1 |
| Analyte or Component Names | Biperiden hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H29 N O . Cl H |
| Synonym | Biperiden hydrochloride, (1 RS)-1-[(1 RS,2 SR,4 RS)-Bicyclo[2.2.1 ]hept-5-en-2-yl]-1-phenyl-3-(piperidin-1-yl)propan-1-ol hydrochloride, Akineton hydrochloride, Akinophyl, Biperidene hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride, alpha-(Bicyclo[2.2.1 ]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.O[C@](CCN1CCCCC1)([C@H]2 C[C@H]3 C[C@@H]2 C=C3)c4ccccc4 |
| IUPAC Name | (1 R)-1-[(1 R,4 R,5 S)-5-bicyclo[2.2.1 ]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |