Learn More
ACROBiosystems Biotinylated SARS-CoV-2 S protein (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F), His, Avitag
Brand:  ACROBiosystems SPN-C82E6-200ug
Encompass_Preferred
 View more versions of this product
Product Code. 17206570
  • 2770.52€ / 200µg
Estimated Shipment: 03-02-2025
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.8 kDa.
Specifications
Specifications
QHD43416.1
200 μg
Val 16 - Pro 1213
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19
Precision Avi
His Tag & Avi Tag
Powder
139.8 kDa
Biotinylated SARS-CoV-2 S protein, His,Avitag (SPN-C82E6) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F, R683A, R685A,F817P, A892P, A899P, A942P, K986P, V987P)). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively.)
-20°C
S protein
SARS-CoV-2
HEK293
MABSol™ Biotin Labeled Proteins
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.