Learn More
ACROBiosystems Biotinylated Human TROP-2 / TACSTD2 Protein, His, Avitag™

Description
TROP-2 is a single-copy gene in human cells, and encodes a type-1 transmembrane glycoprotein which is over-expressed in various malignancies, also referred to as tumor associated calcium signal transducer 2 (TACSTD2), GA733-1 or M1S1. TROP-2 is related to epithelial cell adhesion molecule (EpCAM), also called TROP-1, gp40, and KSA. Trop-1 and Trop-2 are homologous to serum IGF-II-binding proteins and appear as signal transducers. Thus, they likely represent novel cell-surface receptors and may play a role in regulating the growth of carcinoma cells.
Specifications
Specifications
| Accession Number | NP_002344.2 |
| Molecular Weight (g/mol) | 31.6 kDa |
| Quantity | 25 μg |
| Source | Biotinylated Human TROP-2, His,Avitag (TR2-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1). |
| Immunogen | His 27 - Thr 274 |
| Storage Requirements | -20°C |
| Gene Alias | TACSTD2,GA733-1,M1S1,TROP2 |
| Common Name | TROP-2 |
| Conjugate | Precision Avi |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.