missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The NPTX1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human NPTX1. This antibody recognizes human, mouse, rat antigen. The NPTX1 antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications
Specifications
| Antigen | NPTX1 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | Biotin |
| Gene Symbols | DKFZp686J2446; MGC105123; NP1 |
| Host Species | Rabbit |
| Immunogen | Peptide |
| Purification Method | Antigen affinity purification |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?