missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Biotin Antibody (rBTN/8819) [DyLight 405], Novus Biologicals™

Product Code. 30063244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30063244 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30063244 Supplier Novus Biologicals™ Supplier No. NBP327043V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Biotin Monoclonal antibody specifically detects Biotin in Nonspecies specific samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),In-situ Hybridization,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biotin
Applications Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Immunocytochemistry
Classification Monoclonal
Clone rBTN/8819
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias Vitamin B4
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.