missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BICD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 474.00€
Specifications
| Antigen | BICD2 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677240
|
Novus Biologicals
NBP2-91999-0.02ml |
0.02 mL |
204.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661051
|
Novus Biologicals
NBP2-91999-0.1ml |
0.1 mL |
474.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BICD2 Polyclonal antibody specifically detects BICD2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BICD2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 23299 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bicaudal D homolog 2 (Drosophila), KIAA0699 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 440-620 of human BICD2 (NP_001003800.1). HVAVAEAGELREQLKALRSTHEAREAQHAEEKGRYEAEGQALTEKVSLLEKASRQDRELLARLEKELKKVSDVAGETQGSLSVAQDELVTFSEELANLYHHVCMCNNETPNRVMLDYYREGQGGAGRTSPGGRTSPEARGRRSPILLPKGLLAPEAGRADGGTGDSSPSPGSSLPSPLSDP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title