missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate, 98+%, Thermo Scientific™

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

31.60€ - 167.00€

Chemical Identifiers

CAS 41859-67-0
Molecular Formula C19H20ClNO4
Molecular Weight (g/mol) 361.82
MDL Number MFCD00078970
InChI Key IIBYAHWJQTYFKB-UHFFFAOYSA-N
Synonym bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
PubChem CID 39042
ChEBI CHEBI:47612
IUPAC Name 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
SMILES CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464019
SDS View Documents
Thermo Scientific Alfa Aesar
J61412.ME
500mg
31.60€
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15444019
SDS View Documents
Thermo Scientific Alfa Aesar
J61412.03
1g
48.50€
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15454019
SDS View Documents
Thermo Scientific Alfa Aesar
J61412.06
5g
167.00€
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41859-67-0
361.82
IIBYAHWJQTYFKB-UHFFFAOYSA-N
39042
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
C19H20ClNO4
MFCD00078970
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CHEBI:47612
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

41859-67-0
MFCD00078970
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
361.82
CHEBI:47612
≥98%
C19H20ClNO4
14,1195
IIBYAHWJQTYFKB-UHFFFAOYSA-N
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
39042
361.82
Bezafibrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 255-567-9

missing translation for 'rtecsNumber' : UE8755000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

Product Identifier
  • Bezafibrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Bezafibrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.