missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate, 98+%, Thermo Scientific™

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

Brand:  Thermo Scientific Alfa Aesar J61412.06

Quantity 5g

 View more versions of this product

Product Code. 15454019

  • 167.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

41859-67-0
361.82
IIBYAHWJQTYFKB-UHFFFAOYSA-N
39042
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
C19H20ClNO4
MFCD00078970
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CHEBI:47612
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

41859-67-0
MFCD00078970
14,1195
IIBYAHWJQTYFKB-UHFFFAOYSA-N
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
39042
361.82
Bezafibrate
C19H20ClNO4
5g
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
361.82
CHEBI:47612
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.