missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate, 98+%, Thermo Scientific™

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

Brand:  Thermo Scientific Alfa Aesar J61412.03


Quantity 1g

 View more versions of this product

Product Code. 15444019

  • 48.50€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

41859-67-0
361.82
IIBYAHWJQTYFKB-UHFFFAOYSA-N
39042
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
C19H20ClNO4
MFCD00078970
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CHEBI:47612
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

41859-67-0
MFCD00078970
14,1195
IIBYAHWJQTYFKB-UHFFFAOYSA-N
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
39042
361.82
Bezafibrate
C19H20ClNO4
1g
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
361.82
CHEBI:47612
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bezafibrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Bezafibrate

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.