missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol Hydrochloride, TRC

Product Code. 17971675
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971675 10 mg 10mg
30136489 100 mg 100mg
30137093 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971675 Supplier Toronto Research Chemicals Supplier No. TRCB32798010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63659-19-8
Formula Weight 343.1914
InChI Formula InChI=1S/C18H29NO3.ClH/c1-14(2)19-11-17(20)13-22-18-7-5-15(6-8-18)9-10-21-12-16-3-4-16;/h5-8,14,16-17,19-20H,3-4,9-13H2,1-2H3;1H
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
Molecular Formula C18 H29 N O3 . Cl H
Molecular Weight (g/mol) 343.89
SMILES Cl.CC(C)NCC(O)COc1ccc(CCOCC2CC2)cc1
Synonym Betaxolol Hydrochloride, (2RS)-1-[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]-3-[(1-methylethyl)amino]propan-2-ol hydrochloride, 2-Propanol, 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-[(1-methylethyl)amino]-, hydrochloride (1:1), 2-Propanol, 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-[(1-methylethyl)amino]-, hydrochloride (9CI), Betapress, Betoptic, Betoptima, Kerlone, Optipress, SL 75212, SLD 212
Chemical Name or Material Betaxolol Hydrochloride

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.