Learn More
Beta-Tubulin III Mouse, anti-Human, Unconjugated, Clone: TUJ1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60052
This item is not returnable.
View return policy
Description
- The TUJ1 antibody reacts with beta-tubulin III, an ∼50 - 55 kDa structural protein that is a component of tubulin
- Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits
- The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types
- Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation
- It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells
- The TUJ1 antibody is expected to recognize all mammalian homologs of beta-tubulin III
Specifications
| Beta-Tubulin III | |
| Monoclonal | |
| Unconjugated | |
| 250 μL | |
| Human, Mouse, Rat, Other | |
| Neural Cells, PSC-Derived, Neurons |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| TUJ1 | |
| Mouse | |
| Primary | |
| IgG2a κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.