missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Tubulin III Mouse, anti-Human, Alexa Fluor 488, Clone: AA10, STEMCELL Technologies
Description
- The AA10 antibody reacts with beta-tubulin III, an ∼50 - 55 kDa structural protein that is a constituent of tubulin
- Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits
- The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types
- Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation
- It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells
- The AA10 antibody is expected to recognize all mammalian homologs of beta-tubulin III
Specifications
Specifications
| Antigen | Beta-Tubulin III |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | AA10 |
| Conjugate | Alexa Fluor 488 |
| Host Species | Mouse |
| Quantity | 25 μg |
| Primary or Secondary | Primary |
| Target Species | Human, Rat, Mouse |
| Isotype | IgG2a κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?