missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (TUBB/4470), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20754PE
This item is not returnable.
View return policy
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| beta Tubulin | |
| Monoclonal | |
| PE | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| Recombinant full-length human beta Tubulin protein (TUBB5) | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 203068 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| TUBB/4470 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction