missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-endorphin Mouse anti-Human, Clone-CLIP/1449, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54324-100UG
This item is not returnable.
View return policy
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Beta-endorphin | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| CLIP/1449 | |
| Unconjugated | |
| PBS with No Preservative | |
| POMC | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| 100 μg | |
| Neuroscience, Nutrient Sensing in the Brain | |
| 5443 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction