missing translation for 'onlineSavingsMsg'
Learn More

Abcam Beta-Elemene, Anticancer Agent, 204.35 Da, 25 mg

Product Code. 32109562
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109562 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109562 Supplier Abcam Supplier No. ab14517125mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anticancer agent that induces cell cycle arrest and apoptosis.

  • Molecular weight: 204.35 Da
  • Anticancer agent: Induces G2M cell cycle arrest and apoptosis
  • Mechanism: Induces caspase-3, -7 and -9 activities; decreases Bcl-2 expression; induces mitochondrial release of cytochrome c; inhibits microtubule polymerization; downregulates uPA, uPAR, MMP-2, and MMP-9 expression
  • Effects: Shows antiproliferative and antitumor effects
  • Composition: 100% β-Elemene

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% beta-elemene
Content And Storage Store at 4°C.
Shipping Condition Ambient
Target Molecule Beta-Elemene
Formulation 100% β-Elemene
Quantity 25 mg
For Use With (Application) Anticancer research, Apoptosis study, Cell cycle analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.