Learn More
NucA nuclease Antibody, Clone: 1028937, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB100635-SP
 View more versions of this product
Product Code. 18794577
  • 201.00€ / 25µg
Description
NucA Nuclease Monoclonal antibody specifically detects NucA Nuclease in samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NucA Nuclease
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
DcD
E. coli-derived Benzonase Nuclease/NucA, Met1-Asn266, Accession # P13717
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Purified
ELISA
1028937
ELISA
P13717
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
233874471
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.