Learn More
Benzo[a]pyrene-3,6-quinone, TRC
Product Code. 18040530
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18040530 1 mg 1mg
17969739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18040530 Supplier Toronto Research Chemicals Supplier No. TRCB2058651MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3067-14-9
Molecular Formula C20 H10 O2
Molecular Weight (g/mol) 282.29
Synonym Benzo[a]pyrene-3,6-dione, 3,6-Dihydrobenzo[a]pyrene-3,6-dione
IUPAC Name benzo[a]pyrene-3,6-dione
SMILES O=C1C=Cc2ccc3c4ccccc4C(=O)c5ccc1c2c35
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C20H10O2/c21-17-10-6-11-5-7-13-12-3-1-2-4-14(12)20(22)16-9-8-15(17)18(11)19(13)16/h1-10H
Formula Weight 282.0681
Chemical Name or Material Benzo[a]pyrene-3,6-quinone