CAS RN 3067-14-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzo[a]pyrene-3,6-quinone, TRC
CAS: 3067-14-9 Molecular Formula: C20 H10 O2 Molecular Weight (g/mol): 282.29 Synonym: Benzo[a]pyrene-3,6-dione,3,6-Dihydrobenzo[a]pyrene-3,6-dione IUPAC Name: benzo[a]pyrene-3,6-dione SMILES: O=C1C=Cc2ccc3c4ccccc4C(=O)c5ccc1c2c35