Learn More
Benzhydrazide, 98%
Product Code. p-7082033
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11314599 25 g 25g
11414983 100 g 100g
11424983 500 g 500g
3 options
Product Code. 11314599
Brand: Thermo Scientific Alfa Aesar A12268.14
Request Bulk or Custom Format
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 613-94-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007596
InChI Key WARCRYXKINZHGQ-UHFFFAOYSA-N
Synonym benzoylhydrazine, benzhydrazide, benzoic hydrazide, benzoyl hydrazine, benzoic acid, hydrazide, benzoic acid hydrazide, benzohydrazine, benzoylhydrazide, benzoyl hydrazide, hydrazine, benzoyl
PubChem CID 11955
ChEBI CHEBI:38454
IUPAC Name benzohydrazide
SMILES NNC(=O)C1=CC=CC=C1
Specifications
Melting Point 113°C to 117°C
Quantity 25 g
Beilstein 471797
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material Benzhydrazide