Learn More
Benzenes Mixture for HJ 584-2010 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30318692
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318692 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30318692 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000554ME
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 71-43-2, 100-41-4, 98-82-8, 108-38-3, 95-47-6, 106-42-3
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol