Learn More
Bel-Art™ SP Scienceware™ Glove Box Clamping Rings
Product Code. 11775084 Shop All SP Scienceware Products
Click to view available options
For Use With (Equipment):
Fits 6 in. Circular Port Holes
Fits 8 in. Circular Port Holes
Unit Size:
Each
Pack of 2
Product Code. 11775084
Brand: Bel-Art™ H500290200
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Adjustable rings hold gloves in place
  • Fit Techni-Dome™ glove boxes(Mfr. No. 500300000) or other glove boxes with circular port holes of the correct diameter
Specifications
For Use With (Equipment) Fits 6 in. Circular Port Holes
Product Type Clamping Ring
For Use With (Application) Fasten Glove Firmly to Glove Box