Learn More
Behenamide, TRC
Product Code. 30137197
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137197 5 g 5g
30137071 25 g 25g
17939364 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137197 Supplier Toronto Research Chemicals Supplier No. TRCB1311505G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3061-75-4
Molecular Formula C22H45NO
Molecular Weight (g/mol) 339.6
Synonym Docosanamide, Alflow B 10, BNT, BNT 11, BNT 22, BNT 22H, Behenamide, Behenic Acid Amide, Behenic Amide, Behenylamide, Crodamide BR, Denon LS 8, Diamid B 45, Diamid B 90, Diamid BL, Diamid BM, Finawax B, Finawax BR, Incroslip B, Kemamide B, Lutamide BR;
IUPAC Name docosanamide
SMILES NC(CCCCCCCCCCCCCCCCCCCCC)=O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C22H45NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H2,23,24)
Formula Weight 339.3501
Chemical Name or Material Behenamide