CAS RN 3061-75-4
IUPAC Name:
docosanamide
Synonyms:
behenamide
Molecular Weight (g/mol):
339.61
Molecular Formula:
C22H45NO
InChi Key:
ORAWFNKFUWGRJG-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCC(N)=O
1
–
1
of
1
results
1
Behenamide, TRC
CAS: 3061-75-4 Molecular Formula: C22H45NO Molecular Weight (g/mol): 339.6 Synonym: Docosanamide,Alflow B 10,BNT,BNT 11,BNT 22,BNT 22H,Behenamide,Behenic Acid Amide,Behenic Amide,Behenylamide,Crodamide BR,Denon LS 8,Diamid B 45,Diamid B 90,Diamid BL,Diamid BM,Finawax B,Finawax BR,Incroslip B,Kemamide B,Lutamide BR; IUPAC Name: docosanamide SMILES: NC(CCCCCCCCCCCCCCCCCCCCC)=O