Learn More
BD Vacutainer™ Tube Holder
Product Code. 12507876
Change view
Click to view available options
Size:
Standard
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12507876 Standard Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12507876 Supplier BD Supplier No. 364815
  • ONLINE EXCLUSIVE PRICE
    11.85€
    10.90€ / Pack of 250
    Save 0.95€
    8% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Low-cost, single-use holder compliant with OSHA regulations
  • Compatible with the entire BD Vacutainer system of blood collection products
  • Standard size, clear
  • Non-sterile
  • Clear
  • Single use, non-stackable
  • Latex free
Compliance
Complies with OSHA Safety and Health Information Bulletin (SHIB) on the disposal of contaminated needles and blood tube holders used for phlebotomy
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Disposable Single-use
For Use With (Application) Reduces the hazards of contact with Blood borne pathogens compared to reusable holders
Latex Free Yes
Size Standard
Product Type Tube Holder