Learn More
BD Vacutainer™ Pronto™ Quick Release Needle Holder
Click to view available options
Material:
Plastic
Unit Size:
Pack of 20
Description
- Reusable plastic holder (100 estimated uses per holder), designed for use with all BD Vacutainer single and multiple sample needles, blood collection sets and Luer adapter
- Designed to prevent overtightening when needle is secured and to facilitate a smooth and trouble-free release
- Requires no assembly and can be used with any sharps container
Specifications
Specifications
| For Use With (Application) | Prevent over-tightening when needle is secured |
| For Use With (Equipment) | BD Vacutainer single and multiple sample needles, blood collection sets and luer adapter |
| Material | Plastic |
| Product Type | Quick Release Needle Holder |