missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Mouse nterleukin-12 (IL-12), p40

Product Code. 15847798
Click to view available options
Quantity:
2 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15847798

Brand: BD 554594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Determined by SDS-PAGE, and an absorption assay based on the Beer-Lambert law

  • Storage Buffer: Frozen aqueous buffered solution containing BSA and glycerol

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 40μg/mL
For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Name Mouse IL-12 Recombinant Protein
Purification Method SDS-PAGE
Quantity 2 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.