missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Mouse Interleukin-13 (IL-13)
Click to view available options
Quantity:
5 μg
Unit Size:
Each
Description
Frozen aqueous buffered solution containing 1.0mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Mouse
- Storage Buffer: Frozen aqueous buffered solution containing BSA
- Activity Assay: Measured by proliferation using TF-1 indicator cells
- Specific Activity Range: 0.6 - 5 x 107 Units/mg
- ED50: 0.2 to 1.5ng/ml ; Observed dose response relationship: 50 fold
Bioassay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Name | Mouse IL-13 Recombinant Protein |
| Purification Method | SDS-PAGE |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Form | Solution |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction