missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Mouse IL-1β

Product Code. 15857788
Click to view available options
Quantity:
5 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15857788

Brand: BD 554577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Molecular weight: 17 KDa
  • Reactivity: Mouse
  • Contains 153 amino acid residues
  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA
  • Form: Lyophilized
  • >95% pure

Avoid multiple freeze-thaws of product.

Bioassay, ELISA Standard

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioassay, ELISA Standard
Formulation Lyophilized in an aqueous Buffered solution containing BSA
Molecular Weight (g/mol) 17
Name Mouse IL-1 beta Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Lyophilized

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.