missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Interleukin-8 (IL-8)
Description
Lyophilized aqueous buffered solution containing 50μg bovine serum albumin per 1μg recombinant human IL-8 with no preservatives
- Reactivity: Human
- Storage Buffer: Lyophilized in an aqueous buffered solution containing BSA
Enzyme-linked immunosorbent assay standard, functional assay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | ELISA, Functional Assay |
| Formulation | Lyophilized in an aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 8 |
| Name | Human IL-8 Recombinant Protein |
| Quantity | 20 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Lyophilized |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?