missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Interleukin-4 (IL-4)
Click to view available options
Quantity:
5 μg
Unit Size:
Each
Description
Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution, pH 7.2, and containing biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity: The activity range is measured in a proliferation assay using TF-1 indicator cells
- Specific Activity: 0.4 - 5.0 × 108 Units/mg
- ED50: 20 - 250pg/ml
- Observed dose response relationship: 100 fold
Enzyme-linked immunosorbent assay, bioassay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 15 |
| Name | Human IL-4 Recombinant Protein |
| pH Range | 7.2 |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction