missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD P-selectin - IgG Fusion Protein
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
Description
- Composed of the signal sequence, the lectin domain, the EGF-like repeat, and the first two complement-binding domains of mouse P-selectin (CD62P) fused to the Fc region (hinge, CH2 and CH3) of human IgG1
- Binds to P-selectin ligand expressed on mouse neutrophils and myeloid cell lines
Immunoprecipitation, Flow Cytometry
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | Flow Cytometry, Immunoprecipitation |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Mouse P-selectin IgG Fusion Protein |
| Purification Method | Affinity Chromatography |
| Quantity | 0.1 mg |
| Storage Requirements | -20°C |
| Cross Reactivity | Mouse |
| Form | Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction