missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ V-C-N Inhibitor (Thayer and Martin)

Product Code. 17342301
Click to view available options
:
V-C-N Inhibitor (Thayer and Martin); BBL; 10mL
This item is not returnable. View return policy

Product Code. 17342301

Brand: BD 212228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For selection of pathogenic Neisseria, Thayer-Martin Agar and Transgrow medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Additive VCN Inhibitor
Form Lyophilized
For Use With (Application) Selection of pathogenic Neisseria, Thayer-Martin Agar and Transgrow medium
Includes 300μg Vancomycin, 750μg Colistin, 1200U Nystatin
Packaging Type Bottle
Quantity 10 mL
compliance-icons
Product Identifier
  • Vial V-C-N Inhibitor 10ml 10 Ea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : colistin sulphate, nystatin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.