Learn More
BD BBL™ Middlebrook OADC Enrichment
Product Code. 11837463
Click to view available options
Quantity:
500 mL
10 x 20 mL
Packaging Type:
Bottle
Tube
Unit Size:
200mL
500mL
Product Code. 11837463
Brand: BD 212351
Edge
Used as a growth supplement for microbiology research. BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.
  • Dehydrated format
  • Culture media
  • DNR (Latex) free
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Middlebrook OADC Enrichment
Form Viscous
For Use With (Application) Dehydrated culture media
Packaging Type Bottle
Quantity 500 mL