Learn More
BD BBL™/Difco™ TB Fluorescent Stain Kit T
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
Includes
One 250mL bottle each: Auramine –Rhodamine T, Decolorizer TM and Potassium Permanganate
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
Specifications
| Type | TB Fluorescent Stain Kit |
| Quantity | Each |
| Includes | Auramine Rhodamine T, decolorizer TM, and potassium permanganate |
| Format | Bottle |
| For Use With (Application) | For staining mycobacteria by the Truant, Brett and Thomas fluorescent procedure |
| Storage Requirements | 15° to 30°C |
| Packaging Type | Bottle |
| Stain Type | Fluorescent Stain Set T (Auramine Rhodamine T, Decolorizer TM and Potassium Permanganate) |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Flammable liquid Category 3
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P235-Keep cool.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P403-Store in a well-ventilated place.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
For in vitro Diagnostic Use.