missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Supplement B with Reconstituting Fluid
Click to view available options
:
Supplement B; BD Difco; 10mL
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Additive | Supplement B |
| Form | Liquid |
| For Use With (Application) | Formulation of culture media for the cultivation of heterotrophic microorganisms |
| Packaging Type | Vial |
| Quantity | 10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction