missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCS1L Antibody (9G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42922
This item is not returnable.
View return policy
Description
BCS1L Monoclonal antibody specifically detects BCS1L in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| BCS1L | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 9G10 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| BCS, BCS1, BCS1 (yeast homolog)-like, BCS1-like (S. cerevisiae), BCS1-like (yeast), BCS1-like protein, BJSGRACILE, FLNMS, h-BCS, h-BCS1, mitochondrial chaperone BCS1, mitochondrial complex III assembly, PTD | |
| BCS1-like(yeast) - (Uniprot# Q9Y276) | |
| 100 μg | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 617 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction