missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Bcl-6 |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL14612 |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P41182, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?