missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bax Monoclonal antibody specifically detects Bax in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
Specifications
| Antigen | Bax |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 6A7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated to peptides corresponding to amino acids 3-16 of murine Bax (CGSGEQLGSGGPTSS), amino acids 3-16 of human Bax (CGSGEQPRGGGPTSS), and amino acids 12-24 shared by Bax from both species (CGPTSSEQIMKTGA). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?