missing translation for 'onlineSavingsMsg'
Learn More

Bax Antibody (6A7) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18606776 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18606776 0.025 mg 25µg
18614328 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606776 Supplier Novus Biologicals Supplier No. NBP3089850.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Bax Monoclonal antibody specifically detects Bax in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bax
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 6A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4
Host Species Rabbit
Immunogen KLH conjugated to peptides corresponding to amino acids 3-16 of murine Bax (CGSGEQLGSGGPTSS), amino acids 3-16 of human Bax (CGSGEQPRGGGPTSS), and amino acids 12-24 shared by Bax from both species (CGPTSSEQIMKTGA).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 581
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.