missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€ - 507.00€
Specifications
| Antigen | BATF |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620459
|
Novus Biologicals
NBP2-41191-0.025mg |
0.025 mg |
178.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18147937
|
Novus Biologicals
NBP2-41191 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BATF Polyclonal specifically detects BATF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| BATF | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 10538 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BATF. The immunogen is located within the first 50 amino acids of BATF. Amino Acid Squence: SFSRSPPPGKQDSSDD | |
| Primary | |
| Multiple isoforms of BATF are known to exist. Despite its predicted molecular weight, BATF often migrates at a higher than expected molecular weight. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
| BATF | |
| IgG | |
| Affinity Purified | |
| 14 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title