missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bat Immunoglobulin Antibody (BT1-4F10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80590
This item is not returnable.
View return policy
Description
Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Bat Immunoglobulin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 0.5 μg/mL-1.0 μg/mL, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence : 1:100-1:1000, Immunoprecipitation | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Microchiropteran bats (confirmed against big brown, little brown, Red, Silverhair, Hoary bats) and porcine. Does not react with Megachiropteran bats (confirmed against P. poliocephalus, P. giganteus, P. rodricensis, R. aegyptiacus, P. vampyrus, E. helvum, P.pumilus, P. hypomelanus), human, mouse, rat, hamster, canine, feline, guinea pig, and bovine. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| BT1-4F10 | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| Microchiropteran bat (Eptesicus fuscus) spleen cells. | |
| 0.1 mL | |
| Primary | |
| Bat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction