Learn More
Bassoon Antibody (SAP7F407), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB120-13249AF532
Product Code. 18729601
  • 944.00€ / 0.20mL
Description
Bassoon Monoclonal antibody specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)