missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BANF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€
Specifications
| Antigen | BANF1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
BANF1 Polyclonal specifically detects BANF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
| BANF1 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| BAFMGC111161, barrier to autointegration factor 1, barrier-to-autointegration factor, BCRG1, BCRP1, Breakpoint cluster region protein 1, D14S1460 | |
| BANF1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAH05942 | |
| 8815 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human BANF1. The immunogen is located within the last 50 amino acids of BANF1. Amino Acid Squence: LKKDEDLFREWLKDT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title