missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Band 3/AE 1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81066-1-RR-20UL
This item is not returnable.
View return policy
Description
The Band 3/AE 1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Band 3/AE 1. This antibody recognizes human antigen. The Band 3/AE 1 antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Specifications
| Band 3/AE 1 | |
| Recombinant Monoclonal | |
| 800 μg/mL | |
| AE1; BND3; CD233; DI; EMPB3; EPB3; FR; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SW; WD; WD1; WR | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Recombinant Antibody | |
| IgG |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 3K21 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6521 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction