missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baculovirus Envelope gp64 protein Antibody (AcV5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00436-500UG
This item is not returnable.
View return policy
Description
Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Western Blot.
Specifications
| Baculovirus Envelope gp64 protein | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| The AcV5 antibody reacts with the gp64 envelope protein of the baculovirus Autographa californica (AcMNPV). | |
| Store at 4C. Do not freeze. |
| Western Blot | |
| AcV5 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| The immunogen for this antibody is baculovirus envelope gp64 protein. | |
| 500 μg | |
| Primary | |
| Virus | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction