missing translation for 'onlineSavingsMsg'
Learn More

Baclofen Ethyl Ester Hydrochloride, Mikromol™

Product Code. 30052276
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30052276

Brand: Mikromol MM1235.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Baclofen Ethyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Baclofen Ethyl Ester Hydrochloride
CAS 204503-99-1
Analyte or Component Names Baclofen Ethyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H16 Cl N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym beta-(Aminomethyl)-4-chlorobenzenepropanoic Acid Ethyl Ester Hydrochloride (1:1), Benzenepropanoic acid, beta-(aminomethyl)-4-chloro-, ethyl ester, hydrochloride (9CI), Ethyl beta-(aminomethyl)-4-chlorobenzenepropanoate hydrochloride (1:1)
InChI Formula InChI=1S/C12H16ClNO2.ClH/c1-2-16-12(15)7-10(8-14)9-3-5-11(13)6-4-9;/h3-6,10H,2,7-8,14H2,1H3;1H
SMILES Cl.CCOC(=O)CC(CN)c1ccc(Cl)cc1
IUPAC Name ethyl 4-amino-3-(4-chlorophenyl)butanoate;hydrochloride
Molecular Weight (g/mol) 278.17
Quantity 25 mg
Formula Weight 277.0636 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.