missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH2 Antibody (PCRP-BACH2-5B11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44619-100ug
This item is not returnable.
View return policy
Description
BACH2 Monoclonal antibody specifically detects BACH2 in Human samples. It is validated for Flow Cytometry
Specifications
| BACH2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 60468 | |
| Store at 4⪚C. | |
| IgG2a κ |
| Flow Cytometry | |
| PCRP-BACH2-5B11 | |
| Unconjugated | |
| BTB and CNC homolog 2, BTB and CNC homology 1, basic leucine zipper transcription factor 2, transcription regulator protein BACH2 | |
| Recombinant fragment (around aa11-132) of human BACH2 protein (Uniprot: Q9BYV9). Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction