missing translation for 'onlineSavingsMsg'
Learn More

B7-H3/CD276 Antibody (6A1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37657-0.025ml

 View more versions of this product

Product Code. 18668605

  • 217.00€ / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B7-H3/CD276
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
CD276
Purified recombinant fragment of human CD276 expressed in E. Coli.
Unpurified
RUO
Primary
Human
Ascites
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
6A1
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Mouse
57 kDa
0.025 mL
Immunology
80381
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only