missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Raf Antibody (CL4004) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59035AF750
This item is not returnable.
View return policy
Description
B-Raf Monoclonal antibody specifically detects B-Raf in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| B-Raf | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
| This antibody was developed using a recombinant protein derived from P15056, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Phospho Specific | |
| 673 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4004 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction