missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam B-Phycoerythrin Conjugation Kit, Lightning-Link™, 60 μg
Description
- Labeling time: < 4 hrs
- Hands-on time: 30 secs
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Label or Dye | B-Phycoerythrin |
| Shipping Condition | Ambient |
| Target Molecule | Antibody |
| Detection Method | Fluorescence |
| Quantity | 60 μg |
| For Use With (Application) | Antibody labeling |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?